missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thenoin, 97%, Thermo Scientific™

Product Code. 11480528
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27761-02-0
Molecular Formula C10H8O2S2
Molecular Weight (g/mol) 224.292
MDL Number MFCD00022505
InChI Key OGWZIOZTPNLTCR-UHFFFAOYSA-N
Synonym 2,2'-thenoin, 2-hydroxy-1,2-di-2-thienylethanone, 2-hydroxy-1,2-di 2-thienyl ethan-1-one, 2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one, 2-hydroxy-1,2-di thiophen-2-yl ethanone, 2-hydroxy-1,2-bis thiophen-2-yl ethanone, maybridge1_002119, 1,2-dithien-2-ylhydroxyethanone, 2-hydroxy-1,2-dithien-2-ylethanone, alpha-hydroxy-2-thenyl 2-thienyl ketone
PubChem CID 2734800
IUPAC Name 2-hydroxy-1,2-dithiophen-2-ylethanone
SMILES C1=CSC(=C1)C(C(=O)C2=CC=CS2)O

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Formula Weight 224.29
Percent Purity 97%
Chemical Name or Material 2,2'-Thenoin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'