Learn More
2-[(2-Phenoxyethyl)thio]acetic acid, 97%, Thermo Scientific™
38.60 € - 192.00 €
Chemical Identifiers
CAS | 75434-70-7 |
---|---|
Molecular Formula | C10H12O3S |
Molecular Weight (g/mol) | 212.263 |
MDL Number | MFCD00085152 |
InChI Key | SHWKIHWUFNTMIB-UHFFFAOYSA-N |
Synonym | 2-2-phenoxyethyl thio acetic acid, 2-2-phenoxyethyl sulfanyl acetic acid, 2-phenoxyethyl sulfanyl acetic acid, maybridge1_005030, 2-2-phenoxyethylthio acetic acid, 2-phenoxy-ethylsulfanyl acetic acid, 2-phenoxy-ethylsulfanyl-acetic acid, 2-2-phenoxyethylsulfanyl acetic acid |
PubChem CID | 2779087 |
IUPAC Name | 2-(2-phenoxyethylsulfanyl)acetic acid |
SMILES | C1=CC=C(C=C1)OCCSCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10039391
|
Thermo Scientific Maybridge
KM06649DA |
1 g |
38.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10540922
|
Thermo Scientific Maybridge
KM06649EA |
10 g |
98.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10052074
|
Thermo Scientific Maybridge
KM06649EB |
25 g |
192.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
75434-70-7 | |
212.263 | |
SHWKIHWUFNTMIB-UHFFFAOYSA-N | |
2779087 | |
C1=CC=C(C=C1)OCCSCC(=O)O |
C10H12O3S | |
MFCD00085152 | |
2-2-phenoxyethyl thio acetic acid, 2-2-phenoxyethyl sulfanyl acetic acid, 2-phenoxyethyl sulfanyl acetic acid, maybridge1_005030, 2-2-phenoxyethylthio acetic acid, 2-phenoxy-ethylsulfanyl acetic acid, 2-phenoxy-ethylsulfanyl-acetic acid, 2-2-phenoxyethylsulfanyl acetic acid | |
2-(2-phenoxyethylsulfanyl)acetic acid |
Specifications
75434-70-7 | |
97% | |
C10H12O3S | |
MFCD00085152 | |
SHWKIHWUFNTMIB-UHFFFAOYSA-N | |
2-(2-phenoxyethylsulfanyl)acetic acid | |
2779087 | |
97% |
97.0 | |
Amber Glass Bottle | |
C6H5O(CH2)2SCH2CO2H | |
2-2-phenoxyethyl thio acetic acid, 2-2-phenoxyethyl sulfanyl acetic acid, 2-phenoxyethyl sulfanyl acetic acid, maybridge1_005030, 2-2-phenoxyethylthio acetic acid, 2-phenoxy-ethylsulfanyl acetic acid, 2-phenoxy-ethylsulfanyl-acetic acid, 2-2-phenoxyethylsulfanyl acetic acid | |
C1=CC=C(C=C1)OCCSCC(=O)O | |
212.263 | |
212.27 | |
2-[(2-Phenoxyethyl)thio]acetic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.