missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenoxy)-N-methylethylamine, 96%, Thermo Scientific™

221.64 €

Chemical Identifiers

CAS 72955-82-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD07400597
InChI Key VOIPEKOXTFLQNY-UHFFFAOYSA-N
Synonym 2-2-methoxyphenoxy-n-methylethanamine, 2-2-methoxyphenoxy-n-methylethylamine, 2-2-methoxyphenoxy ethyl methyl amine, 2-2-methoxy-phenoxy-ethyl-methyl-amine, 2-2-methoxyphenoxy ethyl methylamine, 2-2-methoxyphenoxy-n-methyl-ethanamine, 2-2-methoxyphenoxy ;-n-methylethanamine, ethanamine,2-2-methoxyphenoxy-n-methyl
PubChem CID 6485441
IUPAC Name 2-(2-methoxyphenoxy)-N-methylethanamine
SMILES CNCCOC1=CC=CC=C1OC
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11449450
View Documents
Thermo Scientific Alfa Aesar
H50299.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

72955-82-9
181.235
VOIPEKOXTFLQNY-UHFFFAOYSA-N
6485441
CNCCOC1=CC=CC=C1OC
C10H15NO2
MFCD07400597
2-2-methoxyphenoxy-n-methylethanamine, 2-2-methoxyphenoxy-n-methylethylamine, 2-2-methoxyphenoxy ethyl methyl amine, 2-2-methoxy-phenoxy-ethyl-methyl-amine, 2-2-methoxyphenoxy ethyl methylamine, 2-2-methoxyphenoxy-n-methyl-ethanamine, 2-2-methoxyphenoxy ;-n-methylethanamine, ethanamine,2-2-methoxyphenoxy-n-methyl
2-(2-methoxyphenoxy)-N-methylethanamine

Specifications

72955-82-9
MFCD07400597
VOIPEKOXTFLQNY-UHFFFAOYSA-N
2-(2-methoxyphenoxy)-N-methylethanamine
6485441
96%
C10H15NO2
2-2-methoxyphenoxy-n-methylethanamine, 2-2-methoxyphenoxy-n-methylethylamine, 2-2-methoxyphenoxy ethyl methyl amine, 2-2-methoxy-phenoxy-ethyl-methyl-amine, 2-2-methoxyphenoxy ethyl methylamine, 2-2-methoxyphenoxy-n-methyl-ethanamine, 2-2-methoxyphenoxy ;-n-methylethanamine, ethanamine,2-2-methoxyphenoxy-n-methyl
CNCCOC1=CC=CC=C1OC
181.235
181.24
2-(2-Methoxyphenoxy)-N-methylethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.