missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenoxy)ethylamine, 97%, Thermo Scientific™

Product Code. 11444850
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120351-90-8
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.172
MDL Number MFCD00235182
InChI Key LXOQYJMQPRGSTL-UHFFFAOYSA-N
Synonym 2-2-fluorophenoxy ethanamine, 2-2-fluorophenoxy ethylamine, 2-2-fluorophenoxy-1-ethanamine, 2-2-fluoro-phenoxy-ethylamine, ethanamine, 2-2-fluorophenoxy, 1-2-aminoethoxy-2-fluorobenzene, 2-2-fluorophenoxy ethyl amine hydrochloride, pubchem19159, acmc-1ci3e, 2-2-fluorophenoxy ;ethanamine
PubChem CID 2106899
IUPAC Name 2-(2-fluorophenoxy)ethanamine
SMILES C1=CC=C(C(=C1)OCCN)F

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 155.17
Percent Purity 97%
Chemical Name or Material 2-(2-Fluorophenoxy)ethylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'