missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
26.20 € - 244.00 €
Chemical Identifiers
CAS | 24650-42-8 |
---|---|
Molecular Formula | C16H16O3 |
Molecular Weight (g/mol) | 256.30 |
MDL Number | MFCD00008475 |
InChI Key | KWVGIHKZDCUPEU-UHFFFAOYSA-N |
Synonym | 2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621 |
PubChem CID | 90571 |
IUPAC Name | 2,2-dimethoxy-1,2-diphenylethanone |
SMILES | COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10242652
|
Thermo Scientific Acros
187840050 |
5 g | Glass bottle |
26.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184010
|
Thermo Scientific Acros
187840500 |
50 g | Glass bottle |
74.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543361
|
Thermo Scientific Acros
187842500 |
250 g | Glass bottle |
244.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24650-42-8 | |
256.30 | |
KWVGIHKZDCUPEU-UHFFFAOYSA-N | |
90571 | |
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
C16H16O3 | |
MFCD00008475 | |
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621 | |
2,2-dimethoxy-1,2-diphenylethanone |
Specifications
24650-42-8 | |
100.0 | |
White to Yellow | |
Authentic | |
C16H16O3 | |
MFCD00008475 | |
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621 | |
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
256.30 | |
256.29 | |
Crystalline Powder |
98.5 | |
64°C to 68°C | |
0.2% max. (75°C, 2 hrs) | |
98.5% min. (GC) | |
C6H5COC(OCH3)2C6H5 | |
14, 3392 | |
KWVGIHKZDCUPEU-UHFFFAOYSA-N | |
2,2-dimethoxy-1,2-diphenylethanone | |
90571 | |
99% | |
2, 2-Dimethoxy-2-phenylacetophenone, 99% |
Safety and Handling
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 246-386-6
missing translation for 'rtecsNumber' : KM5775658
missing translation for 'tsca' : TSCA
RUO – Research Use Only