missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoroacetophenone, 95%, Thermo Scientific Chemicals

Product Code. 11359959
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,2-Difluoroacetophenone acts as an intermediate in the preparation of various substances such as pharmaceuticals. Further, it is used in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides. In addition to this, it undergoes reduction with (-)-diisopinocampheylchloroborane chloride to get R-alcohols of purity 85% ee.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroacetophenone acts as an intermediate in the preparation of various substances such as pharmaceuticals. Further, it is used in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides. In addition to this, it undergoes reduction with (-)-diisopinocampheylchloroborane chloride to get R-alcohols of purity 85% ee.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 395-01-7
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD06246879
InChI Key OLYKCPDTXVZOQF-UHFFFAOYSA-N
Synonym 2,2-difluoroacetophenone, alpha,alpha-difluoroacetophenone, 2,2-difluoro-1-phenyl-ethanone, 2,2-difluoro-1-phenylethan-1-one, a,a-difluoroacetophenone, ethanone, 2,2-difluoro-1-phenyl, difluoromethyl phenyl ketone, difluoroacetophenone, acmc-20aojw, acetophenone, 2,2-difluoro
PubChem CID 273286
IUPAC Name 2,2-difluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)F

Specifications

Melting Point 48°C to 50°C
Boiling Point 53°C to 54°C (12 mmHg)
Odor Characteristic
Refractive Index 1.4955
Quantity 250 mg
Solubility Information Immiscible with water.
Formula Weight 156.13
Percent Purity 95%
Chemical Name or Material 2,2-Difluoroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.