Learn More
2,2-Diethoxyacetophenone, 96%, Thermo Scientific Chemicals
21.00 € - 111.00 €
Chemical Identifiers
CAS | 6175-45-7 |
---|---|
Molecular Formula | C12H16O3 |
Molecular Weight (g/mol) | 208.26 |
MDL Number | MFCD00009659 |
InChI Key | PIZHFBODNLEQBL-UHFFFAOYSA-N |
Synonym | 2,2-diethoxyacetophenone, ethanone, 2,2-diethoxy-1-phenyl, phenylglyoxal diethyl acetal, 2,2-diethoxy-1-phenylethan-1-one, alpha,alpha-diethoxyacetophenone, unii-w4bcz1mak3, 2,2-diethoxy acetophenone, ccris 5222, benzoylformaldehyde diethyl acetal, glyoxal, phenyl-, 2-diethyl acetal |
PubChem CID | 22555 |
IUPAC Name | 2,2-diethoxy-1-phenylethanone |
SMILES | CCOC(OCC)C(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743401
|
Thermo Scientific Acros
210620050 |
5 g | Glass bottle |
21.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10592991
|
Thermo Scientific Acros
210621000 |
100 g | Glass bottle |
111.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6175-45-7 | |
208.26 | |
PIZHFBODNLEQBL-UHFFFAOYSA-N | |
22555 | |
CCOC(OCC)C(=O)C1=CC=CC=C1 |
C12H16O3 | |
MFCD00009659 | |
2,2-diethoxyacetophenone, ethanone, 2,2-diethoxy-1-phenyl, phenylglyoxal diethyl acetal, 2,2-diethoxy-1-phenylethan-1-one, alpha,alpha-diethoxyacetophenone, unii-w4bcz1mak3, 2,2-diethoxy acetophenone, ccris 5222, benzoylformaldehyde diethyl acetal, glyoxal, phenyl-, 2-diethyl acetal | |
2,2-diethoxy-1-phenylethanone |
Specifications
6175-45-7 | |
100.0 | |
1.0300g/mL | |
128°C | |
95% min. (GC) | |
C6H5COCH(OC2H5)2 | |
MFCD00009659 | |
1.03 | |
Solubility in water: 0.16g/L (25°C) | |
CCOC(OCC)C(=O)C1=CC=CC=C1 | |
208.26 | |
7 mPa.s (20°C) | |
96% | |
2, 2-Diethoxyacetophenone, 98% |
97.5 | |
Salmon to Yellow | |
131°C to 134°C (10.0 mmHg) | |
Authentic | |
C12H16O3 | |
1.4985 to 1.5005 | |
07, I, 361 | |
2,2-diethoxyacetophenone, ethanone, 2,2-diethoxy-1-phenyl, phenylglyoxal diethyl acetal, 2,2-diethoxy-1-phenylethan-1-one, alpha,alpha-diethoxyacetophenone, unii-w4bcz1mak3, 2,2-diethoxy acetophenone, ccris 5222, benzoylformaldehyde diethyl acetal, glyoxal, phenyl-, 2-diethyl acetal | |
PIZHFBODNLEQBL-UHFFFAOYSA-N | |
2,2-diethoxy-1-phenylethanone | |
22555 | |
208.26 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-220-4
missing translation for 'rtecsNumber' : MD3284000
missing translation for 'tsca' : TSCA
- 2,2-Diethoxyacetophenone
RUO – Research Use Only