missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethoxy)anisole, 97+%, Thermo Scientific™

Product Code. 15439077
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4463-59-6
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
MDL Number MFCD00017892
InChI Key PBRPKYRJVDJZTF-UHFFFAOYSA-N
Synonym 2-2-bromoethoxy anisole, 1-2-bromoethoxy-2-methoxybenzene, 2-2-bromoethoxy-1-methoxybenzene, 2-bromoethyl 2-methoxyphenyl ether, 2-2-methoxyphenoxy ethylbromide, pubchem3133, 2-methoxyphenoxy ethylbromide, ksc495i5p, akos bc-2659, timtec-bb sbb008602
PubChem CID 1799099
IUPAC Name 1-(2-bromoethoxy)-2-methoxybenzene
SMILES COC1=CC=CC=C1OCCBr

Specifications

Quantity 5 g
Formula Weight 231.09
Percent Purity ≥97%
Chemical Name or Material 2-(2-Bromoethoxy)anisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'