missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, for biochemistry
79.50 € - 257.28 €
Chemical Identifiers
CAS | 6976-37-0 |
---|---|
Molecular Formula | C8H19NO5 |
Molecular Weight (g/mol) | 209.24 |
MDL Number | MFCD00002853 |
InChI Key | OWMVSZAMULFTJU-UHFFFAOYSA-N |
Synonym | bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane |
PubChem CID | 81462 |
ChEBI | CHEBI:41250 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(CO)N(CCO)C(CO)(CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10797582
|
Thermo Scientific Chemicals
327721000 |
100 g |
79.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10711274
|
Thermo Scientific Chemicals
327725000 |
500 g |
257.28 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
6976-37-0 | |
209.24 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
C8H19NO5 | |
MFCD00002853 | |
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
Specifications
6976-37-0 | |
99 | |
103.0°C to 104.0°C | |
8.8 to 11.0 (1% aq. soln. at 25°C) | |
99+% | |
C8H19NO5 | |
MFCD00002853 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.24 | |
CHEBI:41250 | |
99+% | |
Solution |
(0.1M in H2O), (0.1 M in H2O) at 0nm, 0.02 max. at 340nm, 0.04 max. at 280nm | |
100 | |
Yellow to Orange | |
Authentic | |
Plastic bottle | |
(HOCH2CH2)2NC(CH2OH)3 | |
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.24 | |
Biochemistry | |
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
missing translation for 'einecsNumber' : 230-237-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only