missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethylcyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 11464577
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2408-37-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00045550
InChI Key ZPVOLGVTNLDBFI-UHFFFAOYSA-N
Synonym 2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s
PubChem CID 17000
IUPAC Name 2,2,6-trimethylcyclohexan-1-one
SMILES CC1CCCC(C1=O)(C)C

Specifications

Density 0.904
Boiling Point 178°C to 179°C
Flash Point 51°C (125°F)
Refractive Index 1.447
Quantity 5 g
UN Number UN1224
Beilstein 1905593
Solubility Information Slightly miscible with water.
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material 2,2,6-Trimethylcyclohexanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'