missing translation for 'onlineSavingsMsg'
Learn More

2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

29.00 € - 243.00 €

Chemical Identifiers

CAS 92028-57-4
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD01125264
InChI Key ZLYUUANOICYAAL-UHFFFAOYSA-N
Synonym 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 292956
IUPAC Name 2-(2,5-dimethylpyrrol-1-yl)benzoic acid
SMILES CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10780924
View Documents
Thermo Scientific Maybridge
CC39901CB
250 mg
29.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10421313
View Documents
Thermo Scientific Maybridge
CC39901DA
1 g
75.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
10133964
View Documents
Thermo Scientific Maybridge
CC39901DE
5 g
243.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

92028-57-4
215.25
ZLYUUANOICYAAL-UHFFFAOYSA-N
292956
CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O
C13H13NO2
MFCD01125264
2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
2-(2,5-dimethylpyrrol-1-yl)benzoic acid

Specifications

92028-57-4
100.0
Amber Glass Bottle
MFCD01125264
ZLYUUANOICYAAL-UHFFFAOYSA-N
2-(2,5-dimethylpyrrol-1-yl)benzoic acid
292956
97%
96.5
97%
C13H13NO2
2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O
215.25
215.25
2-(2, 5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.