missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%, Thermo Scientific Chemicals

Product Code. 10771062
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2154-67-8
Molecular Formula C9H14NO3
Molecular Weight (g/mol) 184.22
MDL Number MFCD00041847
Synonym 3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 202°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 184.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
Show More Show Less
Product Identifier
  • 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.