missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%, Thermo Scientific Chemicals

Product Code. 10455851
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2154-67-8
Molecular Formula C9H14NO3
Molecular Weight (g/mol) 184.22
MDL Number MFCD00041847
Synonym 3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 202°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 184.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
Show More Show Less
Product Identifier
  • 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'