missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethylpentane, 98%, Thermo Scientific Chemicals

Product Code. 11400207
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-87-7
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00060873
InChI Key GUMULFRCHLJNDY-UHFFFAOYSA-N
PubChem CID 14058
IUPAC Name 2,2,4,4-tetramethylpentane
SMILES CC(C)(C)CC(C)(C)C

Specifications

Density 0.72
Boiling Point 121°C
Flash Point 6°C (43°F)
Odor Petroleum-like
Linear Formula (CH3)3CCH2C(CH3)3
Quantity 50 g
UN Number UN3295
Synonym di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl
Formula Weight 128.26
Percent Purity 98%
Chemical Name or Material 2,2,4,4-Tetramethylpentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'