Learn More
2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals
31.05 € - 236.18 €
Chemical Identifiers
CAS | 434-45-7 |
---|---|
Molecular Formula | C8H5F3O |
Molecular Weight (g/mol) | 174.122 |
MDL Number | MFCD00000420 |
InChI Key | KZJRKRQSDZGHEC-UHFFFAOYSA-N |
Synonym | trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 |
PubChem CID | 9905 |
IUPAC Name | 2,2,2-trifluoro-1-phenylethanone |
SMILES | C1=CC=C(C=C1)C(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494130
|
Thermo Scientific Alfa Aesar
A11403.06 |
5 g |
31.05 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400243
|
Thermo Scientific Alfa Aesar
A11403.14 |
25 g |
84.62 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410243
|
Thermo Scientific Alfa Aesar
A11403.22 |
100 g |
236.18 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
434-45-7 | |
174.122 | |
KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
9905 | |
C1=CC=C(C=C1)C(=O)C(F)(F)F |
C8H5F3O | |
MFCD00000420 | |
trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
2,2,2-trifluoro-1-phenylethanone |
Specifications
434-45-7 | |
1.24 | |
42°C (108°F) | |
1.4595 | |
UN1224 | |
trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
C1=CC=C(C=C1)C(=O)C(F)(F)F | |
174.122 | |
174.12 | |
2,2,2-Trifluoroacetophenone |
-40°C | |
165°C to 166°C | |
C8H5F3O | |
MFCD00000420 | |
1866286 | |
KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-phenylethanone | |
9905 | |
98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 207-103-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only