missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals

28.44 € - 268.34 €

Chemical Identifiers

CAS 75-80-9
Molecular Formula C2H3Br3O
Molecular Weight (g/mol) 282.757
MDL Number MFCD00004671
InChI Key YFDSDPIBEUFTMI-UHFFFAOYSA-N
Synonym tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
PubChem CID 6400
IUPAC Name 2,2,2-tribromoethanol
SMILES C(C(Br)(Br)Br)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11412988
View Documents
Thermo Scientific Alfa Aesar
A18706.06
5 g
28.44 €
5g
Please to purchase this item. Need a web account? Register with us today!
11422988
View Documents
Thermo Scientific Alfa Aesar
A18706.14
25 g
91.35 €
25g
Please to purchase this item. Need a web account? Register with us today!
11432988
View Documents
Thermo Scientific Alfa Aesar
A18706.22
100 g
268.34 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

Solubility
Soluble in alcohol, ether, benzene and warmpetroleum ether. Partially soluble in water.

Notes
Light Sensitive. Store in cool and well ventilated area. Keep container tightly closed.
Specifications

Chemical Identifiers

75-80-9
282.757
YFDSDPIBEUFTMI-UHFFFAOYSA-N
6400
C(C(Br)(Br)Br)O
C2H3Br3O
MFCD00004671
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
2,2,2-tribromoethanol

Specifications

75-80-9
92°C to 93°C (10 mmHg)
MFCD00004671
Light sensitive
Soluble in alcohol,ether,benzene and warmpetroleum ether. Partially soluble in water.
C(C(Br)(Br)Br)O
282.757
282.76
2,2,2-Tribromoethanol
77°C to 80°C
C2H3Br3O
1733249
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
YFDSDPIBEUFTMI-UHFFFAOYSA-N
2,2,2-tribromoethanol
6400
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 200-903-1

missing translation for 'rtecsNumber' : KM3675000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only