missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(1H-Tetrazol-5-yl)phenol, 97%

Product Code. 11352749
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51449-77-5
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.152
MDL Number MFCD09037787
InChI Key SQPPXOJPOBGAAU-UHFFFAOYSA-N
Synonym phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol
PubChem CID 42553059
IUPAC Name 6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C2NNN=N2)C(=O)C=C1

Specifications

Melting Point ∽230°C (decomposition)
Quantity 1 g
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 2-(1H-Tetrazol-5-yl)phenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.