missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 11496350
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-41-2
Molecular Formula C15H23BN2O2
Molecular Weight (g/mol) 274.17
MDL Number MFCD06798280
InChI Key RKTXCUGUPDEZHR-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-pyrrolidin-1-yl pyridine-3-boronic acid pinacol ester, 2-1-pyrrolidinyl pyridine-3-boronic acid pinacol ester, 2-pyrrolidin-1-ylpyridine-3-boronic acid, pinacol ester, 2-pyrrolidinopyridine-3-boronic acid, pinacol ester, 2-1-pyrrolidino pyridine-3-boronic acid pinacol ester, 2-pyrrolidin-1-yl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-1-pyrrolidinyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 44755171
IUPAC Name 2-pyrrolidin-1-yl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(N=CC=C1)N1CCCC1

Specifications

Melting Point 76°C to 78°C
Quantity 1 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 274.17
Percent Purity 95%
Chemical Name or Material 2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'