missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Naphthyl)ethanoyl chloride, 97%, Thermo Scientific™
24.20 € - 167.00 €
Chemical Identifiers
CAS | 5121-00-6 |
---|---|
Molecular Formula | C12H9ClO |
Molecular Weight (g/mol) | 204.65 |
MDL Number | MFCD00175852 |
InChI Key | DSVAZLXLRDXHKO-UHFFFAOYSA-N |
Synonym | 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride |
PubChem CID | 2795386 |
IUPAC Name | 2-naphthalen-1-ylacetyl chloride |
SMILES | ClC(=O)CC1=CC=CC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10591883
|
Thermo Scientific Maybridge
S07397CB |
250 mg |
24.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10724773
|
Thermo Scientific Maybridge
S07397DA |
1 g |
51.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10265594
|
Thermo Scientific Maybridge
S07397EA |
10 g |
167.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
5121-00-6 | |
204.65 | |
DSVAZLXLRDXHKO-UHFFFAOYSA-N | |
2795386 | |
ClC(=O)CC1=CC=CC2=CC=CC=C12 |
C12H9ClO | |
MFCD00175852 | |
2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride | |
2-naphthalen-1-ylacetyl chloride |
Specifications
5121-00-6 | |
Amber Glass Bottle | |
MFCD00175852 | |
DSVAZLXLRDXHKO-UHFFFAOYSA-N | |
2-naphthalen-1-ylacetyl chloride | |
2795386 | |
95% |
95% | |
C12H9ClO | |
2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride | |
ClC(=O)CC1=CC=CC2=CC=CC=C12 | |
204.65 | |
204.66 | |
2-(1-Naphthyl)ethanoyl chloride |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-(1-Naphthyl)ethanoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.