missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3,4-Oxadiazol-2-yl)phenol, 98%, Thermo Scientific™

Product Code. 11325459
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-65-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00020713
InChI Key HKVNQGYCUGIYTA-UHFFFAOYSA-N
Synonym 2-1,3,4-oxadiazol-2-yl phenol, fenadiazole, 2-1,3,4-oxadiazol-2-yl benzenol, oinxxhyenyvypb-uhfffaoysa-n, acmc-1bzdh, ortho-1,3,4-oxadiazol-2-yl-phenol, 6-3h-1,3,4-oxadiazol-2-ylidene cyclohexa-2,4-dien-1-one
PubChem CID 6738087
IUPAC Name 6-(3H-1,3,4-oxadiazol-2-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C2NN=CO2)C(=O)C=C1

Specifications

Quantity 1 g
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 2-(1,3,4-Oxadiazol-2-yl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'