Learn More
(1R)-(+)-Camphor, 98%, Thermo Scientific Chemicals
25.30 € - 184.00 €
Chemical Identifiers
CAS | 464-49-3 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.24 |
MDL Number | MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149 |
InChI Key | DSSYKIVIOFKYAU-XVKPBYJWSA-N |
Synonym | +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone |
PubChem CID | 159055 |
ChEBI | CHEBI:15396 |
IUPAC Name | (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
SMILES | CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414290
|
Thermo Scientific Alfa Aesar
A10708.14 |
25 g |
25.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424290
|
Thermo Scientific Alfa Aesar
A10708.22 |
100 g |
57.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434290
|
Thermo Scientific Alfa Aesar
A10708.36 |
500 g |
184.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
Solubility
Soluble in water (0.1 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
464-49-3 | |
152.24 | |
DSSYKIVIOFKYAU-XVKPBYJWSA-N | |
159055 | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
C10H16O | |
MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149 | |
+-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone | |
CHEBI:15396 | |
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2 |
Specifications
464-49-3 | |
0.99 | |
65°C (149°F) | |
C10H16O | |
UN2717 | |
14,1732 | |
Soluble in water (0.1g/L at 20°C). | |
+43° (c=10 in ethanol) | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one | |
159055 | |
152.24 | |
(1R)- (+)-Camphor |
175°C to 180°C | |
204°C | |
Camphor-like | |
MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149 | |
2042745 | |
+-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone | |
DSSYKIVIOFKYAU-XVKPBYJWSA-N | |
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2 | |
152.24 | |
CHEBI:15396 | |
98% |
Safety and Handling
GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335-H500
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
missing translation for 'einecsNumber' : 207-355-2
missing translation for 'rtecsNumber' : EX1260000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only