missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 11336976
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303180-11-2
Molecular Formula C5H6N2O5
Molecular Weight (g/mol) 174.112
MDL Number MFCD00149323
InChI Key GLINCONFUZIMCN-UHFFFAOYSA-N
Synonym 1h-pyrazole-3,5-dicarboxylic acid hydrate, 3,5-pyrazoledicarboxylic acid monohydrate, 1h-pyrazole-3,5-dicarboxylic acid monohydrate, pyrazole-3,5-dicarboxylic acid, hydrate, acmc-1aezk, 3,5-pyrazoledicarboxylic acid hydrate, 1h-pyrazole-3,5-dicarboxylicacidmonohydrate, 1h-pyrazole-3,5-dicarboxylic acid; hydrate, 1h-pyrazole-3,5-dicarboxylic acid hydrate 1:1
PubChem CID 2723723
IUPAC Name 1H-pyrazole-3,5-dicarboxylic acid;hydrate
SMILES C1=C(NN=C1C(=O)O)C(=O)O.O

Specifications

Melting Point ∽294°C (decomposition)
Quantity 25 g
Beilstein 147821
Formula Weight 174.11 (156.09 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole-3,5-dicarboxylic acid monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'