missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Indole-2-methanol, 96%, Thermo Scientific Chemicals
79.40 € - 260.00 €
Chemical Identifiers
CAS | 24621-70-3 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.17 |
InChI Key | XEEANGGQJOWRTG-UHFFFAOYSA-N |
Synonym | 1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol |
PubChem CID | 98783 |
IUPAC Name | 1H-indol-2-ylmethanol |
SMILES | C1=CC=C2C(=C1)C=C(N2)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15582373
|
Thermo Scientific Acros
459810010 |
1 g |
79.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15592373
|
Thermo Scientific Acros
459810050 |
5 g |
260.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24621-70-3 | |
147.17 | |
1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol | |
1H-indol-2-ylmethanol |
C9H9NO | |
XEEANGGQJOWRTG-UHFFFAOYSA-N | |
98783 | |
C1=CC=C2C(=C1)C=C(N2)CO |
Specifications
24621-70-3 | |
Authentic | |
Glass Bottle | |
1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol | |
C1=CC=C2C(=C1)C=C(N2)CO | |
147.17 | |
147.17 | |
1H-Indole-2-methanol |
72.0°C to 78.0°C | |
95% min. (GC) | |
C9H9NO | |
XEEANGGQJOWRTG-UHFFFAOYSA-N | |
1H-indol-2-ylmethanol | |
98783 | |
96% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1H-Indole-2-methanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only