Learn More
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
101.51 € - 379.89 €
Chemical Identifiers
CAS | 51851-37-7 |
---|---|
Molecular Formula | C14H19F13O3Si |
Molecular Weight (g/mol) | 510.367 |
MDL Number | MFCD00042333 |
InChI Key | AVYKQOAMZCAHRG-UHFFFAOYSA-N |
Synonym | 1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane |
PubChem CID | 103991 |
IUPAC Name | triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
SMILES | CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480478
|
Thermo Scientific Alfa Aesar
B24620.06 |
5 g |
101.51 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490478
|
Thermo Scientific Alfa Aesar
B24620.14 |
25 g |
379.89 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.
Solubility
Miscible with alcohol, aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Incompatible with moisture and oxidizing agents.
Chemical Identifiers
51851-37-7 | |
510.367 | |
AVYKQOAMZCAHRG-UHFFFAOYSA-N | |
103991 | |
CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC |
C14H19F13O3Si | |
MFCD00042333 | |
1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane | |
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
Specifications
220°C | |
51851-37-7 | |
CF3(CF2)5(CH2)2Si(OCH2CH3)3 | |
97°C (206°F) | |
Miscible with alcohol,aromatic and aliphatic hydrocarbons. | |
CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC | |
510.367 | |
510.37 | |
1.345 | |
1.333 |
<-38°C | |
C14H19F13O3Si | |
MFCD00042333 | |
1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane | |
AVYKQOAMZCAHRG-UHFFFAOYSA-N | |
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane | |
103991 | |
97% | |
Moisture sensitive | |
1H,1H,2H,2H-Perfluorooctyltriethoxysilane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
missing translation for 'einecsNumber' : 257-473-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.