missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™

37.10 € - 212.00 €

Chemical Identifiers

CAS 101947-16-4
Molecular Formula C16H19F17O3Si
Molecular Weight (g/mol) 610.38
MDL Number MFCD00042334
InChI Key MLXDKRSDUJLNAB-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g
PubChem CID 145374
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329255
View Documents
Thermo Scientific Alfa Aesar
L16585.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11339255
View Documents
Thermo Scientific Alfa Aesar
L16585.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11349255
View Documents
Thermo Scientific Alfa Aesar
L16585.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.

Solubility
Miscible with ethanol and tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

101947-16-4
610.38
MLXDKRSDUJLNAB-UHFFFAOYSA-N
145374
CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
C16H19F17O3Si
MFCD00042334
1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane

Specifications

103°C to 106°C (3mmHg)
C16H19F17O3Si
MFCD00042334
100°C (212°F)
Miscible with ethanol and tetrahydrofuran.
CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
610.38
610.38
1.342
1.41
101947-16-4
CF3(CF2)7(CH2)2Si(OCH2CH3)3
UN1760
1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g
MLXDKRSDUJLNAB-UHFFFAOYSA-N
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
145374
97%
Moisture sensitive
1H,1H,2H,2H-Perfluorodecyltriethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.