missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 12-well Insert 2in1, PET-Membrane
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
285.00 € - 422.00 €
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
16104967
|
BRAND™
782736 |
8 μm | 48 x 1 |
422.00 €
Pack of 48 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
16114967
|
BRAND™
782737 |
8 μm | 4 x 9 |
285.00 €
Pack of 36 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
16184957
|
BRAND™
782730 |
0.4 μm | 48 x 1 |
422.00 €
Pack of 48 |
In Stock
Log in to see stock. |
||||
16194957
|
BRAND™
782731 |
0.4 μm | 4 x 9 |
285.00 €
Pack of 36 |
In Stock
Log in to see stock. |
||||
Description
- Use Inserts in a hanging or standing position
- For 12-well plates or culture dishes
- Achieve uniform and controlled cell growth due to surface treatment, without coatings
Specifications
PET | |
Sterile | |
12 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title