missing translation for 'onlineSavingsMsg'
Learn More

11-Azidoundecyltrimethoxysilane, 94%, Thermo Scientific Chemicals

Product Code. 15457237
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334521-23-2
Molecular Formula C14H31N3O3Si
Molecular Weight (g/mol) 317.51
MDL Number MFCD11982878
InChI Key ACSJXAKDVNDMME-UHFFFAOYSA-N
Synonym 11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide
PubChem CID 46779110
SMILES CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC

Specifications

Boiling Point 100°C to 105°C (0.04mmHg)
Quantity 1 g
Linear Formula N3(CH2)11Si(OCH3)3
IUPAC Name 11-azidoundecyl(trimethoxy)silane
Formula Weight 317.51
Percent Purity 94%
Sensitivity Moisture sensitive
Chemical Name or Material 11-Azidoundecyltrimethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'