missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon 100ML Reagecon B1 Brown Colour Reference Solutionaccording to European Pharmacopoeia
Brand: Reagecon EPB101
100ML Reagecon B1 Brown Colour Reference Solutionaccording to European Pharmacopoeia
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COLOUR REFERENCE SOLUTION B1
Signal Word
- ||Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper sulphate, sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction