missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoyl chloride, 98%, Thermo Scientific Chemicals

109.55 € - 434.16 €

Chemical Identifiers

CAS 38460-95-6
Molecular Formula C11H19ClO
Molecular Weight (g/mol) 202.72
MDL Number MFCD00000772
InChI Key MZFGYVZYLMNXGL-UHFFFAOYSA-N
Synonym 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
PubChem CID 38042
IUPAC Name undec-10-enoyl chloride
SMILES ClC(=O)CCCCCCCCC=C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10042050
View Documents
Thermo Scientific Acros
171581000
100 mL Glass bottle
109.55 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10387970
View Documents
Thermo Scientific Acros
171585000
500 mL Glass bottle
434.16 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38460-95-6
202.72
MZFGYVZYLMNXGL-UHFFFAOYSA-N
38042
ClC(=O)CCCCCCCCC=C
C11H19ClO
MFCD00000772
10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
undec-10-enoyl chloride

Specifications

38460-95-6
100.0
0.9400g/mL
93°C
98%
H2C=CH(CH2)8COCl
MFCD00000772
0.94
MZFGYVZYLMNXGL-UHFFFAOYSA-N
undec-10-enoyl chloride
38042
98%
10-Undecenoyl chloride, 98%
97.5
Yellow
120°C to 122°C (10.0 mmHg)
Authentic
C11H19ClO
1.4532 to 1.4552
02, 459
10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
ClC(=O)CCCCCCCCC=C
202.72
202.72
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 253-951-0

missing translation for 'rtecsNumber' : YQ2991000

missing translation for 'tsca' : TSCA

Product Identifier
  • 10-Undecenoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.