Learn More
10-Isocyanatodecyltrimethoxysilane, 95%, Thermo Scientific™
71.00 € - 218.09 €
Chemical Identifiers
CAS | 1310372-82-7 |
---|---|
Molecular Formula | C14H29NO4Si |
Molecular Weight (g/mol) | 303.474 |
MDL Number | MFCD19105613 |
InChI Key | GUOFKJBNQHCAKJ-UHFFFAOYSA-N |
Synonym | 10-isocyanatodecyl trimethoxysilane |
PubChem CID | 87686085 |
IUPAC Name | 10-isocyanatodecyl(trimethoxy)silane |
SMILES | CO[Si](CCCCCCCCCCN=C=O)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406387
|
Thermo Scientific Alfa Aesar
H33555.MD |
250 mg | N/A | ||||||
15496377
|
Thermo Scientific Alfa Aesar
H33555.03 |
1 g |
218.09 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1310372-82-7 | |
303.474 | |
GUOFKJBNQHCAKJ-UHFFFAOYSA-N | |
87686085 | |
CO[Si](CCCCCCCCCCN=C=O)(OC)OC |
C14H29NO4Si | |
MFCD19105613 | |
10-isocyanatodecyl trimethoxysilane | |
10-isocyanatodecyl(trimethoxy)silane |
Specifications
120°C to 125°C (0.1mmHg) | |
C14H29NO4Si | |
MFCD19105613 | |
10-isocyanatodecyl trimethoxysilane | |
CO[Si](CCCCCCCCCCN=C=O)(OC)OC | |
303.474 | |
303.47 | |
Moisture sensitive |
1310372-82-7 | |
OCN(CH2)10Si(OCH3)3 | |
UN2810 | |
GUOFKJBNQHCAKJ-UHFFFAOYSA-N | |
10-isocyanatodecyl(trimethoxy)silane | |
87686085 | |
95% | |
10-Isocyanatodecyltrimethoxysilane |
Safety and Handling
GHS H Statement
H331-H318-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
Your input is important to us. Please complete this form to provide feedback related to the content on this product.