Learn More
1-Trimethylsilyl-1,4-pentadiyne, 98%, Thermo Scientific Chemicals
37.40 € - 505.00 €
Chemical Identifiers
CAS | 71789-10-1 |
---|---|
Molecular Formula | C8H12Si |
Molecular Weight (g/mol) | 136.27 |
MDL Number | MFCD00066350 |
InChI Key | JKYMNPFNLMDSFG-UHFFFAOYSA-N |
Synonym | 1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl |
PubChem CID | 144510 |
IUPAC Name | trimethyl(penta-1,4-diynyl)silane |
SMILES | C[Si](C)(C)C#CCC#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410248
|
Thermo Scientific Alfa Aesar
H53436.03 |
1 g |
37.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420248
|
Thermo Scientific Alfa Aesar
H53436.06 |
5 g |
128.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430248
|
Thermo Scientific Alfa Aesar
H53436.14 |
25 g |
505.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesIncompatible with strong oxidizing agents and ammonia.
Chemical Identifiers
71789-10-1 | |
136.27 | |
JKYMNPFNLMDSFG-UHFFFAOYSA-N | |
144510 | |
C[Si](C)(C)C#CCC#C |
C8H12Si | |
MFCD00066350 | |
1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl | |
trimethyl(penta-1,4-diynyl)silane |
Specifications
42°C (25mmHg) | |
C8H12Si | |
MFCD00066350 | |
8°C (46°F) | |
JKYMNPFNLMDSFG-UHFFFAOYSA-N | |
trimethyl(penta-1,4-diynyl)silane | |
144510 | |
98% | |
1-Trimethylsilyl-1,4-pentadiyne |
71789-10-1 | |
HC≡CCH2C≡CSi(CH3)3 | |
UN1993 | |
1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl | |
C[Si](C)(C)C#CCC#C | |
136.27 | |
136.27 | |
0.789 |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only