Learn More
1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals
59.09 € - 727.00 €
Chemical Identifiers
CAS | 6651-36-1 |
---|---|
Molecular Formula | C9H18OSi |
Molecular Weight (g/mol) | 170.33 |
MDL Number | MFCD00001541 |
InChI Key | SBEMOANGDSSPJY-UHFFFAOYSA-N |
Synonym | 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane |
PubChem CID | 81161 |
IUPAC Name | cyclohexen-1-yloxy(trimethyl)silane |
SMILES | C[Si](C)(C)OC1=CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433127
|
Thermo Scientific Alfa Aesar
A16099.09 |
10 g |
59.09 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443127
|
Thermo Scientific Alfa Aesar
A16099.18 |
50 g |
196.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453127
|
Thermo Scientific Alfa Aesar
A16099.30 |
250 g | N/A | ||||||
Description
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Chemical Identifiers
6651-36-1 | |
170.33 | |
SBEMOANGDSSPJY-UHFFFAOYSA-N | |
81161 | |
C[Si](C)(C)OC1=CCCCC1 |
C9H18OSi | |
MFCD00001541 | |
1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane | |
cyclohexen-1-yloxy(trimethyl)silane |
Specifications
165°C | |
C9H18OSi | |
UN1993 | |
42°C (107°F) | |
Not miscible or difficult to mix. | |
C[Si](C)(C)OC1=CCCCC1 | |
170.33 | |
170.33 | |
1.447 | |
0.883 |
6651-36-1 | |
MFCD00001541 | |
1859394 | |
1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane | |
SBEMOANGDSSPJY-UHFFFAOYSA-N | |
cyclohexen-1-yloxy(trimethyl)silane | |
81161 | |
98% | |
Moisture sensitive | |
1-(Trimethylsiloxy)cyclohexene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 229-675-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.