Learn More
1-Propylamine, 99+%, Thermo Scientific Chemicals
37.29 € - 115.58 €
Chemical Identifiers
CAS | 107-10-8 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.112 |
MDL Number | MFCD00008205 |
InChI Key | WGYKZJWCGVVSQN-UHFFFAOYSA-N |
Synonym | propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine |
PubChem CID | 7852 |
ChEBI | CHEBI:39870 |
IUPAC Name | propan-1-amine |
SMILES | CCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358068
|
Thermo Scientific Alfa Aesar
036635.AK |
250 mL |
37.29 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368068
|
Thermo Scientific Alfa Aesar
036635.K2 |
1 L |
115.58 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Propylamine is used in chemical analysis and to make other chemicals. It is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Also it can be used as petroleum additive and preservative. It is used in the production of fiber and engineering plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-10-8 | |
59.112 | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
7852 | |
propan-1-amine |
C3H9N | |
MFCD00008205 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
CHEBI:39870 | |
CCCN |
Specifications
107-10-8 | |
0.719 g/mL | |
-12°C (10°F) | |
C3H9N | |
MFCD00008205 | |
1098243 | |
14,7843 | |
Fully miscible with water. Highly soluble in ethanol and acetone,soluble in benzene and chloroform and slightly soluble in carbon tetrachloride. | |
CCCN | |
59.112 | |
CHEBI:39870 | |
≥99% | |
1-Propylamine |
°C to 83°C | |
48°C to 49°C | |
>99+% | |
1.389 | |
UN1277 | |
Air Sensitive | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
propan-1-amine | |
7852 | |
59.11 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H311-H314-H318-H302
P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
H225-H290-H302-H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYLAMINE
missing translation for 'einecsNumber' : 203-462-3
missing translation for 'rtecsNumber' : UH9100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- n-Propylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1-Propanamine
RUO – Research Use Only