missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, purum, ≥99% (GC), Honeywell™
≥99% (GC), Purum
417.89 € - 676.16 €
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15602520
|
Honeywell Research Chemicals
24135-6X1L |
6 x 1 L | Poly Bottle |
417.89 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15692510
|
Honeywell Research Chemicals
24135-4X2.5L |
4 x 2.5 L | Poly Bottle |
676.16 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
-127° | |
0.804g/mL at 25°C | |
0.137 | |
700°F | |
C3H8O | |
MFCD00002941 | |
UN1274 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
≥99% (GC) | |
1-Propanol |
71-23-8 | |
97° | |
22°C (71.6°F) | |
Liquid | |
CH3CH2CH2OH | |
10mmHg (147°C), 14.9mmHg (20°C) | |
1098242 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1g/mol | |
purum |
Safety and Handling
P210-P261-P280-P305 + P351 + P338
H225-H318-H336
missing translation for 'dotInformation' : Class 3 - PG 2 - n-Propanol
missing translation for 'einecsNumber' : 200-746-9
missing translation for 'rtecsNumber' : UH8225000