missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™

Product Code. 11414837
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20109-09-5
Molecular Formula C14H16O
Molecular Weight (g/mol) 200.281
MDL Number MFCD00060883
InChI Key LBEMYSKBSYEVAG-UHFFFAOYSA-N
Synonym 1-phenylethynyl-1-cyclohexanol, 1-phenylethynyl cyclohexanol, 1-2-phenylethynyl cyclohexan-1-ol, 1-phenylethynylcyclohexanol, acmc-1cey1, 1-phenylethynyl-cyclohexanol, 1-phenylethynyl cyclohexanol #, cyclohexanol,1-2-phenylethynyl, 1-2-phenylethynyl-1-cyclohexanol, 1-2-phenyl-ethynyl-cyclohexan-1-ol
PubChem CID 593776
IUPAC Name 1-(2-phenylethynyl)cyclohexan-1-ol
SMILES C1CCC(CC1)(C#CC2=CC=CC=C2)O

Specifications

Melting Point 60°C to 62°C
Quantity 5 g
Formula Weight 200.28
Percent Purity 99%
Chemical Name or Material 1-(Phenylethynyl)cyclohexanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'