missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexanol, 97%, Thermo Scientific Chemicals

53.30 € - 193.00 €

Chemical Identifiers

CAS 1589-60-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00021393
InChI Key DTTDXHDYTWQDCS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
PubChem CID 15319
IUPAC Name 1-phenylcyclohexan-1-ol
SMILES C1CCC(CC1)(C2=CC=CC=C2)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11499896
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20877.09
10 g
53.30 €
10g
Please to purchase this item. Need a web account? Register with us today!
11409906
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20877.18
50 g
193.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1589-60-2
176.259
DTTDXHDYTWQDCS-UHFFFAOYSA-N
15319
C1CCC(CC1)(C2=CC=CC=C2)O
C12H16O
MFCD00021393
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
1-phenylcyclohexan-1-ol

Specifications

1589-60-2
153°C (20 mmHg)
MFCD00021393
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
C1CCC(CC1)(C2=CC=CC=C2)O
176.259
176.26
1-Phenylcyclohexanol
58°C to 62°C
C12H16O
1818360
DTTDXHDYTWQDCS-UHFFFAOYSA-N
1-phenylcyclohexan-1-ol
15319
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 216-456-0

missing translation for 'rtecsNumber' : GW0699900

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only