missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15651370
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -78°C
Color Colorless
Density 0.811g/mL at 25°C
Boiling Point 136°C to 138°C
Flash Point 49°C (120.2°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)4OH
Refractive Index n20/D 1.409(lit.)
Quantity 2.5 L
UN Number UN1105
Beilstein 1730975
Formula Weight 88.15g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material 1-Pentanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'