Learn More
1-Octene, 99+%, Thermo Scientific Chemicals
33.80 € - 305.00 €
Chemical Identifiers
CAS | 111-66-0 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.21 |
MDL Number | MFCD00009548 |
InChI Key | KWKAKUADMBZCLK-UHFFFAOYSA-N |
Synonym | 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene |
PubChem CID | 8125 |
ChEBI | CHEBI:46708 |
IUPAC Name | oct-1-ene |
SMILES | CCCCCCC=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772951
|
Thermo Scientific Acros
301251000 |
100 mL | Glass bottle |
35.70 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10104383
|
Thermo Scientific
301255000 |
500 mL | Glass bottle |
33.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10424035
|
Thermo Scientific Acros
301250010 |
1 L | Glass bottle |
56.80 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10656671
|
Thermo Scientific Acros
301250025 |
2.5 L | Glass bottle |
112.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10093813
|
Thermo Scientific Acros
301250100 |
10 L | Metal Drum |
305.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-66-0 | |
112.21 | |
KWKAKUADMBZCLK-UHFFFAOYSA-N | |
8125 | |
oct-1-ene |
C8H16 | |
MFCD00009548 | |
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene | |
CHEBI:46708 | |
CCCCCCC=C |
Specifications
111-66-0 | |
100.0 | |
0.7150g/mL | |
13°C | |
99% min. (GC) | |
CH3(CH2)5CH=CH2 | |
MFCD00009548 | |
15, 1766 | |
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene | |
KWKAKUADMBZCLK-UHFFFAOYSA-N | |
oct-1-ene | |
8125 | |
0.38 mm2/s (40°C) | |
99+% | |
1-Octene, 99+% |
99.0 | |
-102.0°C | |
121.0°C to 123.0°C | |
Authentic | |
C8H16 | |
1.4080 to 1.4100 | |
01, 221 | |
0.715 | |
Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents | |
CCCCCCC=C | |
112.21 | |
CHEBI:46708 | |
112.21 | |
Liquid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environm
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-893-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only