missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonic Acid Sodium Salt, Ion Pair Chromatography, Solution 0.1M, Fisher Chemical
Click to view available options
Quantity:
250 mL
Packaging:
Amber glass bottle
Specifications
Specifications
Physical Form | Liquid |
Quantity | 250 mL |
MDL Number | 7544 |
Packaging | Amber glass bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'