missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11305379
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5121-00-6
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.65
MDL Number MFCD00175852
InChI Key DSVAZLXLRDXHKO-UHFFFAOYSA-N
Synonym 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride
PubChem CID 2795386
IUPAC Name 2-(naphthalen-1-yl)acetyl chloride
SMILES ClC(=O)CC1=CC=CC2=CC=CC=C12

Specifications

Density 1.23
Boiling Point 130°C (0.6 mmHg)
Odor Pungent
Refractive Index 1.621
Quantity 5 g
UN Number UN3265
Formula Weight 204.66
Percent Purity 95%
Chemical Name or Material 1-Naphthylacetyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'