Learn More
(1-Methylpiperid-3-yl)methylamine, 97%, Thermo Scientific™
75.70 € - 208.00 €
Chemical Identifiers
CAS | 14613-37-7 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.219 |
MDL Number | MFCD06738902 |
InChI Key | KEDTYNCWGSIWBK-UHFFFAOYSA-N |
Synonym | 1-methylpiperidin-3-yl methanamine, 3-aminomethyl-1-methylpiperidine, 1-methyl-3-aminomethyl piperidine, 3-piperidinemethanamine, 1-methyl, c-1-methyl-piperidin-3-yl-methylamine, 1-methyl-3-aminomethyl-piperidine, 1-methyl-3-piperidinemethanamine, 1-1-methylpiperidin-3-yl methanamine, 1-methylpiperid-3-yl methylamine |
PubChem CID | 13675353 |
IUPAC Name | (1-methylpiperidin-3-yl)methanamine |
SMILES | CN1CCCC(C1)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10516154
|
Thermo Scientific Maybridge
CC66113CB |
250 mg | Amber Glass Bottle |
75.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10164104
|
Thermo Scientific Maybridge
CC66113DA |
1 g | Amber Glass Bottle |
208.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
14613-37-7 | |
128.219 | |
KEDTYNCWGSIWBK-UHFFFAOYSA-N | |
13675353 | |
CN1CCCC(C1)CN |
C7H16N2 | |
MFCD06738902 | |
1-methylpiperidin-3-yl methanamine, 3-aminomethyl-1-methylpiperidine, 1-methyl-3-aminomethyl piperidine, 3-piperidinemethanamine, 1-methyl, c-1-methyl-piperidin-3-yl-methylamine, 1-methyl-3-aminomethyl-piperidine, 1-methyl-3-piperidinemethanamine, 1-1-methylpiperidin-3-yl methanamine, 1-methylpiperid-3-yl methylamine | |
(1-methylpiperidin-3-yl)methanamine |
Specifications
14613-37-7 | |
100.0 | |
C7H16N2 | |
1-methylpiperidin-3-yl methanamine, 3-aminomethyl-1-methylpiperidine, 1-methyl-3-aminomethyl piperidine, 3-piperidinemethanamine, 1-methyl, c-1-methyl-piperidin-3-yl-methylamine, 1-methyl-3-aminomethyl-piperidine, 1-methyl-3-piperidinemethanamine, 1-1-methylpiperidin-3-yl methanamine, 1-methylpiperid-3-yl methylamine | |
CN1CCCC(C1)CN | |
128.219 | |
128.22 | |
(1-Methylpiperid-3-yl)methylamine |
96.5 | |
97% | |
MFCD06738902 | |
KEDTYNCWGSIWBK-UHFFFAOYSA-N | |
(1-methylpiperidin-3-yl)methanamine | |
13675353 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger