missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

64.40 € - 174.00 €

Chemical Identifiers

CAS 10250-64-3
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD08271939
InChI Key RTBQZMPYPFQBHA-UHFFFAOYSA-N
PubChem CID 7537625
IUPAC Name 2-methyl-5-phenylpyrazole-3-carboxylic acid
SMILES CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10583874
View Documents
Thermo Scientific Maybridge
CC46601CB
250 mg Amber Glass Bottle
64.40 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10051054
View Documents
Thermo Scientific Maybridge
CC46601DA
1 g Amber Glass Bottle
174.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

10250-64-3
202.213
RTBQZMPYPFQBHA-UHFFFAOYSA-N
2-methyl-5-phenylpyrazole-3-carboxylic acid
C11H10N2O2
MFCD08271939
7537625
CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O

Specifications

10250-64-3
100.0
C11H10N2O2
1-methyl-3-phenyl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-phenyl-2h-pyrazole-3-carboxylic acid, 1-methyl-3-phenylpyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl-3-phenyl, 5-carboxy-1-methyl-3-phenyl-1h-pyrazole, 1-methyl-3-phenyl-1h-5-pyrazolecarboxylic acid, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-phenyl, 1-methyl-3-phenyl-1 h-pyrazole-5-carboxylic acid
CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O
202.213
202.21
1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
96.5
97%
MFCD08271939
RTBQZMPYPFQBHA-UHFFFAOYSA-N
2-methyl-5-phenylpyrazole-3-carboxylic acid
7537625
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.