missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-Pyrrolidinone, 99%, Reagent Grade, Honeywell™

Product Code. 15697960
Click to view available options
Quantity:
1 L
18 L
25 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 8.5 to 10 (at 100.00g/L 20°C)
Melting Point -24°C
Color Colorless
Density 1.028g/mL at 25°C
Boiling Point 204°C (1013 hPa)
Quantity 18 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Odor Amine-like
Packaging Stainless Steel Can
Physical Form Liquid
Vapor Pressure 0.99mmHg (40°C), 0.29mmHg (20°C)
Beilstein 106420
Solubility Information Fully miscible in water
Formula Weight 99.13g/mol
Percent Purity 0.99
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'