missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-Pyrrolidinone, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
1 L
18 L
25 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
pH | 8.5 to 10 (at 100.00g/L 20°C) |
Melting Point | -24°C |
Color | Colorless |
Density | 1.028g/mL at 25°C |
Boiling Point | 204°C (1013 hPa) |
Quantity | 4 x 4 L |
Explosion Limit | 0.095 |
Flash Point | 91°C (195.8°F) |
Ignition Point | 518°F |
Odor | Amine-like |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'