missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride, 90%, Thermo Scientific™

Product Code. 10706295
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-57-6
Molecular Formula C9H8Cl2N2O
Molecular Weight (g/mol) 231.08
MDL Number MFCD09817460
InChI Key SGNORNZSRSQMEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
PubChem CID 24229458
IUPAC Name 1-methylbenzimidazole-5-carbonyl chloride;hydrochloride
SMILES Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 231.08
Percent Purity 90%
Chemical Name or Material 1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'