Learn More
1-Hexylboronic acid, 97%, Thermo Scientific Chemicals
79.50 € - 247.00 €
Chemical Identifiers
CAS | 16343-08-1 |
---|---|
Molecular Formula | C6H15BO2 |
Molecular Weight (g/mol) | 129.994 |
MDL Number | MFCD01074641 |
InChI Key | CXSYDLCMCLCOCA-UHFFFAOYSA-N |
Synonym | n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2 |
PubChem CID | 351064 |
IUPAC Name | hexylboronic acid |
SMILES | B(CCCCCC)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400607
|
Thermo Scientific Alfa Aesar
B22446.88 |
2.5 g |
79.50 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410607
|
Thermo Scientific Alfa Aesar
B22446.09 |
10 g |
247.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16343-08-1 | |
129.994 | |
CXSYDLCMCLCOCA-UHFFFAOYSA-N | |
351064 | |
B(CCCCCC)(O)O |
C6H15BO2 | |
MFCD01074641 | |
n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2 | |
hexylboronic acid |
Specifications
16343-08-1 | |
C6H15BO2 | |
1736080 | |
n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2 | |
B(CCCCCC)(O)O | |
129.994 | |
130 | |
1-Hexylboronic acid |
85°C to 90°C | |
MFCD01074641 | |
Air Sensitive | |
CXSYDLCMCLCOCA-UHFFFAOYSA-N | |
hexylboronic acid | |
351064 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only