missing translation for 'onlineSavingsMsg'
Learn More

1-Heptylamine, 98+%, Thermo Scientific Chemicals

Product Code. 11408750
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-68-2
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008244
InChI Key WJYIASZWHGOTOU-UHFFFAOYSA-N
Synonym heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
PubChem CID 8127
IUPAC Name heptan-1-amine
SMILES CCCCCCCN

Specifications

Melting Point -23°C
Density 0.775
Boiling Point 154°C to 156°C
Flash Point 35°C (95°F)
Odor Amine-like
Linear Formula CH3(CH2)6NH2
Refractive Index 1.4245
Quantity 50 g
UN Number UN2734
Beilstein 1731688
Sensitivity Air Sensitive
Solubility Information Soluble in water 6791mg/L @ 25°C.
Formula Weight 115.22
Percent Purity ≥98%
Chemical Name or Material 1-Heptylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.