missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals

43.00 € - 402.00 €

Chemical Identifiers

CAS 321-38-0
Molecular Formula C10H7F
Molecular Weight (g/mol) 146.164
MDL Number MFCD00003873
InChI Key CWLKTJOTWITYSI-UHFFFAOYSA-N
Synonym fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
PubChem CID 9450
IUPAC Name 1-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A18378.06
5 g
43.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11428307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A18378.14
25 g
137.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11438307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A18378.22
100 g
402.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

321-38-0
146.164
CWLKTJOTWITYSI-UHFFFAOYSA-N
9450
C1=CC=C2C(=C1)C=CC=C2F
C10H7F
MFCD00003873
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
1-fluoronaphthalene

Specifications

321-38-0
1.331
65°C (149°F)
1.593
1906413
Not miscible or difficult to mix in water.
C1=CC=C2C(=C1)C=CC=C2F
146.164
146.17
1-Fluoronaphthalene
-10°C
215°C to 217°C
C10H7F
MFCD00003873
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
CWLKTJOTWITYSI-UHFFFAOYSA-N
1-fluoronaphthalene
9450
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 206-287-0

missing translation for 'rtecsNumber' : QJ7100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only