Learn More
1-Dodecylamine hydrochloride, 97%, Thermo Scientific Chemicals
41.60 € - 142.00 €
Chemical Identifiers
CAS | 929-73-7 |
---|---|
Molecular Formula | C12H28ClN |
Molecular Weight (g/mol) | 221.81 |
MDL Number | MFCD00042017 |
InChI Key | TWFQJFPTTMIETC-UHFFFAOYSA-N |
Synonym | dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd |
PubChem CID | 458426 |
IUPAC Name | dodecan-1-amine;hydrochloride |
SMILES | [Cl-].CCCCCCCCCCCC[NH3+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348735
|
Thermo Scientific Alfa Aesar
L12314.09 |
10 g |
41.60 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358735
|
Thermo Scientific Alfa Aesar
L12314.18 |
50 g |
142.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
929-73-7 | |
221.81 | |
TWFQJFPTTMIETC-UHFFFAOYSA-N | |
458426 | |
[Cl-].CCCCCCCCCCCC[NH3+] |
C12H28ClN | |
MFCD00042017 | |
dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd | |
dodecan-1-amine;hydrochloride |
Specifications
929-73-7 | |
C12H28ClN | |
MFCD00042017 | |
dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd | |
[Cl-].CCCCCCCCCCCC[NH3+] | |
221.81 | |
221.82 | |
1-Dodecylamine hydrochloride |
186°C to 188°C | |
CH3(CH2)11NH2·HCl | |
3553943 | |
TWFQJFPTTMIETC-UHFFFAOYSA-N | |
dodecan-1-amine;hydrochloride | |
458426 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 213-205-7
missing translation for 'rtecsNumber' : JR6750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only