missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

144.00 € - 246.00 €

Chemical Identifiers

CAS 141091-37-4
Molecular Formula C12H21BO2
Molecular Weight (g/mol) 208.11
MDL Number MFCD05663845
InChI Key QNZFUMVTUFOLRT-UHFFFAOYSA-N
Synonym 2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
PubChem CID 10932675
IUPAC Name 2-(cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10402868
View Documents
Thermo Scientific Acros
430315000
500 mg
144.00 €
500mg
Please to purchase this item. Need a web account? Register with us today!
10309827
View Documents
Thermo Scientific Acros
430310010
1 g
246.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141091-37-4
208.11
QNZFUMVTUFOLRT-UHFFFAOYSA-N
10932675
CC1(C)OB(OC1(C)C)C1=CCCCC1
C12H21BO2
MFCD05663845
2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
2-(cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

141091-37-4
100.0
125.0°C (0.2 mmHg)
96% min. (GC)
MFCD05663845
QNZFUMVTUFOLRT-UHFFFAOYSA-N
2-(cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10932675
97%
1-Cyclohexenylboronic acid pinacol ester
96.0
Colorless to Tan
Authentic
C12H21BO2
2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CCCCC1
208.11
208.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 1-Cyclohexenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.